This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01459197_g1
See other CD4 GE Assays ›
Gene Symbol:
Cd4
Gene Name:
Cd4 molecule
Gene Aliases:
W3/25, p55
Chromosome Location:
Chr.4: 157381862 - 157408358 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01459197_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012705.1 NP_036837.1 7 - 8 1328 100
XM_006237331.2 XP_006237393.1 9 - 10 1591 100
XM_006237333.2 XP_006237395.1 7 - 8 1351 100
XM_008763280.1 XP_008761502.1 7 - 8 1308 100
GenBank mRNA M15768.1 - 7 - 8 1328 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400003743.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd4

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
induction by virus of host cell-cell fusion
immune response
cell surface receptor signaling pathway
T cell differentiation
maintenance of protein location in cell
T cell activation
T cell selection
protein palmitoleylation
positive regulation of protein kinase activity
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
positive regulation of calcium-mediated signaling
regulation of T cell activation
positive regulation of T cell activation
glycoprotein binding
zinc ion binding
coreceptor activity
enzyme binding
protein kinase binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400026581.rn.1 TC0400003743.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0