This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01458851_m1
See other S100A16 GE Assays ›
Gene Symbol:
S100a16
Gene Name:
S100 calcium binding protein A16
Gene Aliases:
-
Chromosome Location:
Chr.2: 189923140 - 189928850 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01458851_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108557.2 NP_001102027.1 1 - 2 46 70
GenBank mRNA BC158598.1 - 1 - 2 46 7127572 70
FQ209774.1 - 1 - 2 48 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC158598.1, NM_001108557.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

response to calcium ion
calcium ion binding
protein homodimerization activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200010824.rn.1 TC0200001614.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200010829.rn.1 TC0200001614.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0