This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01455898_m1
See other TCAF1 GE Assays ›
Gene Symbol:
Tcaf1
Gene Name:
TRPM8 channel-associated factor 1
Gene Aliases:
Fam115a, RGD1565474
Chromosome Location:
Chr.4: 72119356 - 72171728 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01455898_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcaf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008762928.1 XP_008761150.1 1 - 2 480 78
XM_342675.6 XP_342676.4 1 - 2 480 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008762928.1, XM_342675.6 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell migration
positive regulation of protein targeting to membrane
positive regulation of anion channel activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0