This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01455569_m1
See other S100A7A GE Assays ›
Gene Symbol:
S100a7a
Gene Name:
S100 calcium binding protein A7A
Gene Aliases:
S100a15
Chromosome Location:
Chr.2: 190058093 - 190062534 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01455569_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a7a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109471.1 NP_001102941.1 1 - 2 103 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109471.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cytokine metabolic process
positive chemotaxis
calcium ion binding
chemoattractant activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200010884.rn.1 TC0200001621.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200010885.rn.1 TC0200001621.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0