This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01450974_g1
See other LIPA GE Assays ›
Gene Symbol:
Lipa
Gene Name:
lipase A, lysosomal acid, cholesterol esterase
Gene Aliases:
Chole, Chole2, LAL, Lip1
Chromosome Location:
Chr.1: 252816536 - 252870302 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01450974_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012732.3 NP_036864.3 5 - 6 818 86
XM_006231275.2 XP_006231337.1 5 - 6 785 86
XM_008760321.1 XP_008758543.1 4 - 5 681 86
GenBank mRNA BC072532.1 - 5 - 6 818 7099460 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100007594.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipa

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytokine production
fatty acid metabolic process
inflammatory response
cell proliferation
lipid catabolic process
sterol metabolic process
lung development
tissue remodeling
homeostasis of number of cells within a tissue
sterol esterase activity
lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100061017.rn.1 TC0100007594.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0