This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01447316_g1
See other ARPP19 GE Assays ›
Gene Symbol:
Arpp19 LOC100360828
Gene Name:
cAMP-regulated phosphoprotein 19
cAMP-regulated phosphoprotein 19-like
Gene Aliases:
Arpp-19
-
Chromosome Location:
Chr.8: 81949582 - 81972054 on Build Rnor_6.0
Chromosome Location:
Chr.8: 119105962 - 119106371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01447316_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arpp19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006243410.2 XP_006243472.1 2 - 3 154 92
GenBank mRNA BC058461.1 - 2 - 3 89 6919944 92
FQ222843.1 - 2 - 3 92 92
Gene Symbol: LOC100360828
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002729972.4 XP_002730018.2 1 80 92

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058461.1,XM_002729972.4 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
mitotic nuclear division
dopamine receptor signaling pathway
negative regulation of catalytic activity
positive regulation of gluconeogenesis
positive regulation of Ras protein signal transduction
regulation of catalytic activity
cell division
protein phosphatase inhibitor activity
receptor binding
protein phosphatase type 2A regulator activity
potassium channel regulator activity
phosphatase inhibitor activity
protein phosphatase 2A binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0