This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01446020_g1
See other CAMP GE Assays ›
Gene Symbol:
Camp
Gene Name:
cathelicidin antimicrobial peptide
Gene Aliases:
CRAMP
Chromosome Location:
Chr.8: 117930557 - 117932371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01446020_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100724.1 NP_001094194.1 1 - 2 205 75
GenBank mRNA AF484553.1 - 1 - 2 190 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003405.rn.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
innate immune response in mucosa
positive regulation of cell proliferation
negative regulation of endopeptidase activity
antibacterial humoral response
negative regulation of growth of symbiont in host
negative regulation of growth of symbiont on or near host surface
positive regulation of angiogenesis
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
killing by host of symbiont cells
cellular response to lipopolysaccharide
cellular response to peptidoglycan
cellular response to interleukin-1
cellular response to interleukin-6
cellular response to tumor necrosis factor
molecular_function
cysteine-type endopeptidase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0