This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01444411_m1
See other BOC GE Assays ›
Gene Symbol:
Boc
Gene Name:
BOC cell adhesion associated, oncogene regulated
Gene Aliases:
-
Chromosome Location:
Chr.11: 61084284 - 61159220 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01444411_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Boc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751052.3 XP_003751100.1 6 - 7 1319 64
XM_006248353.2 XP_006248415.1 6 - 7 1319 64
XM_006248354.2 XP_006248416.1 6 - 7 1319 64
XM_006248355.2 XP_006248417.1 6 - 7 1319 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s)

smoothened signaling pathway
axon guidance
cell projection organization
positive regulation of myoblast differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0