This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01444067_m1
See other CLDND1 GE Assays ›
Gene Symbol:
Cldnd1
Gene Name:
claudin domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 43986352 - 43992739 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01444067_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cldnd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006955.1 NP_001006956.1 3 - 4 503 84
XM_006248204.2 XP_006248266.1 4 - 5 695 84
XM_006248205.2 XP_006248267.1 3 - 4 644 84
XM_006248206.2 XP_006248268.1 4 - 5 546 84
XM_008768633.1 XP_008766855.1 3 - 4 486 84
GenBank mRNA BC083649.1 - 3 - 4 503 7135100 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100001445.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cldnd1

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100008743.rn.1 TC1100001445.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100008744.rn.1 TC1100001445.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0