This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01441384_g1
See other LOC100363500 GE Assays ›
Gene Symbol:
LOC100363500 Creb1
Gene Name:
hypothetical LOC100363500
cAMP responsive element binding protein 1
Gene Aliases:
-
Creb
Chromosome Location:
Chr.18: 25361682 - 25364547 on Build Rnor_6.0
Chromosome Location:
Chr.9: 71229799 - 71298994 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01441384_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100363500
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008772050.1 XP_008770272.1 1 546 77
Gene Symbol: Creb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031017.1 NP_112279.1 6 - 7 592 77
NM_134443.1 NP_604392.1 4 - 5 511 77
XM_006245065.2 XP_006245127.1 6 - 7 598 77
XM_006245066.2 XP_006245128.1 5 - 6 763 77
XM_006245067.2 XP_006245129.1 5 - 6 503 77
XM_006245068.2 XP_006245130.1 4 - 5 450 77
XM_006245069.2 XP_006245131.1 4 - 5 1100 77
XM_008767138.1 XP_008765360.1 3 - 4 2954 77
GenBank mRNA BC097311.1 - 6 - 7 792 77
FQ211925.1 - 6 - 7 657 77
X14788.1 - 6 - 7 592 77
X60002.1 - 4 - 5 511 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LOC100363500
Click here to find siRNAs for Gene Creb1

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein phosphorylation
transforming growth factor beta receptor signaling pathway
axonogenesis
lactation
memory
circadian rhythm
regulation of cell size
response to organic substance
negative regulation of transcription by competitive promoter binding
pituitary gland development
cell differentiation
mammary gland development
positive regulation of transforming growth factor beta3 production
secretory granule organization
response to glucagon
positive regulation of multicellular organism growth
response to drug
regulation of circadian rhythm
regulation of apoptotic process
positive regulation of fat cell differentiation
positive regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hormone secretion
positive regulation of lipid biosynthetic process
protein stabilization
lung epithelium development
lung saccule development
Type I pneumocyte differentiation
cellular response to zinc ion
cellular response to growth factor stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
enzyme binding
histone acetyltransferase binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900006524.rn.1 TC0900000802.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900006525.rn.1 TC0900000802.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0