Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 8 - 9 | 1425 | 60 | |
XM_006256604.2 | XP_006256666.1 | 9 - 10 | 1713 | 60 | ||
XM_008772994.1 | XP_008771216.1 | 9 - 10 | 1553 | 60 | ||
XM_008772995.1 | XP_008771217.1 | 9 - 10 | 2902 | 60 | ||
XM_008772996.1 | XP_008771218.1 | 9 - 10 | 1536 | 60 | ||
XM_008772997.1 | XP_008771219.1 | 8 - 9 | 5178 | 60 | ||
XM_008772998.1 | XP_008771220.1 | 6 - 7 | 710 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
biological_process
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein heterodimerization activity
ureteric bud development
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
biological_process
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042196 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P777777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132845 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042196 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P777777 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P777777 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042196 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042196 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P777777 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P117030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91693 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45534 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9857 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10830746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830747 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830752 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830749 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10830743 | - | - |
Rat Genome 230 2.0 Array | 1382698_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382698_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000007554.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007551.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007552.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007546.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007548.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007555.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007550.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007556.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007557.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007553.rn.1 | TC2000000920.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007549.rn.1 | TC2000000920.rn.1 | RTA TC |
Rat Expression Set 230 | 1382698_at | - | - |
Rat Expression Set 230 | 1382698_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830743 | - | - |
Rat Genome U34 Set | rc_AI011075_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349911 | - | - |
