This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01436995_m1
See other GBA2 GE Assays ›
Gene Symbol:
Gba2
Gene Name:
glucosidase beta 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 59068081 - 59079719 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01436995_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gba2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013091.2 NP_001013109.2 3 - 4 534 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002621.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

glucosylceramide catabolic process
bile acid metabolic process
glycoside catabolic process
central nervous system neuron development
glucosylceramidase activity
beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500019565.rn.1 TC0500002621.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500019568.rn.1 TC0500002621.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0