This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01434919_m1
See other SERPINB1A GE Assays ›
Gene Symbol:
Serpinb1a
Gene Name:
serine (or cysteine) proteinase inhibitor, clade B, member 1a
Gene Aliases:
-
Chromosome Location:
Chr.17: 32903089 - 32911498 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01434919_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serpinb1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031642.1 NP_001026812.1 1 - 2 185 85
GenBank mRNA BC098686.1 - 1 - 2 185 7385870 85

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serpinb1a

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endopeptidase activity
regulation of protein catabolic process
serine-type endopeptidase inhibitor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141420 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023472 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 514083 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 585294 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 441380 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 106371 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 185648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 329632 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 99297 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 116282 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023472 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141420 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141420 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023472 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141420 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023472 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17175 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10794872 - -
Affymetrix Rat Gene 1.0 ST Array 10794869 - -
Affymetrix Rat Gene 1.0 ST Array 10794871 - -
Affymetrix Rat Gene 1.0 ST Array 10794868 - -
Affymetrix Rat Gene 1.0 ST Array 10794867 - -
Affymetrix Rat Gene 1.0 ST Array 10794870 - -
Affymetrix Rat Gene 1.1 ST Array 10794866 - -
Rat Genome 230 2.0 Array 1380346_at - -
Rat Genome 230 2.0 Array 1377034_at - -
Rat Genome 230 2.0 Array 1380346_PM_at - -
Rat Genome 230 2.0 Array 1377034_PM_at - -
Affymetrix HT Rat Focus Array 1380346_at - -
Affymetrix HT Rat Focus Array 1377034_at - -
Affymetrix HT Rat Focus Array 1377034_PM_at - -
Affymetrix HT Rat Focus Array 1380346_PM_at - -
Affymetrix Rat Clariom S Assay PSR1700003617.rn.1 TC1700000506.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700003616.rn.1 TC1700000506.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700003615.rn.1 TC1700000506.rn.1 RTA TC
Rat Expression Set 230 1377034_at - -
Rat Expression Set 230 1377034_PM_at - -
Rat Expression Set 230 1380346_PM_at - -
Rat Expression Set 230 1380346_at - -
Affymetrix Rat Gene 1.0 ST Array 10794866 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21851700 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352285 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0