This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01433623_m1
See other NODAL GE Assays ›
Gene Symbol:
Nodal
Gene Name:
nodal growth differentiation factor
Gene Aliases:
-
Chromosome Location:
Chr.20: 31035729 - 31044096 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01433623_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nodal
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106394.1 NP_001099864.1 1 - 2 481 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000000680.rn.1

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
trophectodermal cell differentiation
trophectodermal cellular morphogenesis
neural fold formation
liver development
placenta development
embryonic placenta development
maternal placenta development
vasculature development
heart looping
inhibition of neuroepithelial cell differentiation
determination of left/right symmetry
gastrulation
nervous system development
brain development
endoderm development
heart development
positive regulation of cell proliferation
embryonic pattern specification
anterior/posterior axis specification
anterior/posterior pattern specification
regulation of signal transduction
polarity specification of proximal/distal axis
regulation of gastrulation
positive regulation of vascular endothelial growth factor production
negative regulation of cell development
positive regulation of pathway-restricted SMAD protein phosphorylation
cell migration
stem cell population maintenance
positive regulation of cell-cell adhesion
lung development
positive regulation of activin receptor signaling pathway
floor plate morphogenesis
embryonic heart tube development
endodermal cell differentiation
nodal signaling pathway
growth
cell migration involved in gastrulation
regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of MAPK cascade
cell fate commitment
negative regulation of cell differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
axial mesodermal cell fate specification
mesendoderm development
cell development
digestive tract morphogenesis
anatomical structure formation involved in morphogenesis
embryonic cranial skeleton morphogenesis
tissue morphogenesis
formation of anatomical boundary
positive regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
digestive system development
embryonic process involved in female pregnancy
maternal process involved in parturition
positive regulation of SMAD protein import into nucleus
SMAD protein signal transduction
left lung morphogenesis
negative regulation of androgen receptor signaling pathway
epiblast cell-extraembryonic ectoderm cell signaling involved in anterior/posterior axis specification
positive regulation of ERK1 and ERK2 cascade
primitive streak formation
transforming growth factor beta receptor signaling pathway involved in primitive streak formation
nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
negative regulation of trophoblast cell migration
negative regulation of chorionic trophoblast cell proliferation
positive regulation of receptor activity
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
receptor agonist activity
type I activin receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000005970.rn.1 TC2000000680.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000005968.rn.1 TC2000000680.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000005969.rn.1 TC2000000680.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0