This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01431782_m1
See other ADAT1 GE Assays ›
Gene Symbol:
Adat1
Gene Name:
adenosine deaminase, tRNA-specific 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 44175865 - 44212101 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01431782_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109611.1 NP_001103081.1 3 - 4 389 94
XM_006255634.2 XP_006255696.1 3 - 4 1229 94
XM_006255635.2 XP_006255697.1 3 - 4 1355 94
XM_006255636.2 XP_006255698.1 3 - 4 1183 94
XM_006255638.1 XP_006255700.1 2 - 3 234 94
XM_008772535.1 XP_008770757.1 3 - 4 1229 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006255638.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
tRNA processing
RNA binding
tRNA-specific adenosine deaminase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1900012244.rn.1 TC1900001484.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0