This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01430677_m1
See other RP2 GE Assays ›
Gene Symbol:
Rp2
Gene Name:
retinitis pigmentosa 2 homolog (human)
Gene Aliases:
RGD1565124, Rp2h
Chromosome Location:
Chr.X: 2066298 - 2116661 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01430677_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109012.1 NP_001102482.1 1 - 2 279 95
XM_008773035.1 XP_008771257.1 1 - 2 279 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0X00001387.rn.1

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
post-Golgi vesicle-mediated transport
cytoskeleton organization
post-chaperonin tubulin folding pathway
biological_process
positive regulation of GTPase activity
molecular_function
actin binding
nucleoside diphosphate kinase activity
GTPase activator activity
ATP binding
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00008844.rn.1 TC0X00001387.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00008840.rn.1 TC0X00001387.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00008843.rn.1 TC0X00001387.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0