This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01429511_m1
See other PCBP3 GE Assays ›
Gene Symbol:
Pcbp3
Gene Name:
poly(rC) binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.20: 12429264 - 12629981 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01429511_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcbp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011945.1 NP_001011945.1 8 - 9 837 95
XM_006256283.2 XP_006256345.1 8 - 9 887 95
XM_006256285.2 XP_006256347.1 5 - 6 471 95
XM_006256286.2 XP_006256348.1 8 - 9 887 95
XM_006256287.2 XP_006256349.1 8 - 9 888 95
XM_006256288.2 XP_006256350.1 8 - 9 885 95
XM_006256289.2 XP_006256351.1 8 - 9 885 95
XM_006256290.2 XP_006256352.1 6 - 7 1886 95
XM_006256291.2 XP_006256353.1 8 - 9 886 95
XM_008772874.1 XP_008771096.1 6 - 7 1907 95
XM_008772875.1 XP_008771097.1 6 - 7 591 95
GenBank mRNA BC079196.1 - 8 - 9 837 7131426 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000000428.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcbp3

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000004024.rn.1 TC2000000428.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0