This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01428418_s1
See other CDKN1A GE Assays ›
Gene Symbol:
Cdkn1a
Gene Name:
cyclin-dependent kinase inhibitor 1A
Gene Aliases:
Cip1, UV96, Waf1
Chromosome Location:
Chr.20: 6348422 - 6358864 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01428418_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdkn1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080782.3 NP_542960.1 3 88
XM_006256128.2 XP_006256190.1 3 1113 88
GenBank mRNA BC100620.1 - 3 7938247 88
U12526.1 - 1 53 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdkn1a

Panther Classification:

Molecular Function -

kinase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
Ras protein signal transduction
regulation of mitotic cell cycle
negative regulation of cell proliferation
response to UV
response to toxic substance
response to organic substance
response to X-ray
response to organonitrogen compound
negative regulation of gene expression
response to organic cyclic compound
negative regulation of cell growth
positive regulation of B cell proliferation
organ regeneration
cellular response to extracellular stimulus
regulation of protein import into nucleus, translocation
cellular response to heat
negative regulation of phosphorylation
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of apoptotic process
positive regulation of programmed cell death
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of protein kinase activity
response to arsenic-containing substance
positive regulation of fibroblast proliferation
response to glucocorticoid
response to corticosterone
regulation of cell cycle
response to hyperoxia
intestinal epithelial cell maturation
cellular response to ionizing radiation
cellular response to UV-B
mitotic cell cycle arrest
cellular senescence
negative regulation of G1/S transition of mitotic cell cycle
regulation of DNA biosynthetic process
positive regulation of reactive oxygen species metabolic process
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
cyclin-dependent protein kinase activating kinase activity
cyclin binding
ubiquitin protein ligase binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000002454.rn.1 TC2000000249.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0