This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01424962_g1
See other LOC100362572 GE Assays ›
Gene Symbol:
LOC100362572
Gene Name:
Mpv17 transgene, kidney disease mutant-like (predicted)-like
Gene Aliases:
-
Chromosome Location:
Chr.10: 3321487 - 3359219 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01424962_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100362572
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008767466.1 XP_008765688.1 3 - 4 481 104
XM_008767468.1 XP_008765690.1 3 - 4 216 104
XM_008767469.1 XP_008765691.1 3 - 4 158 104
XM_008767470.1 XP_008765692.1 3 - 4 245 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of hydrogen peroxide biosynthetic process
reactive oxygen species metabolic process
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
receptor binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20720893 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0