Assay Details
Gene Symbol: LOC100362572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767466.1 | XP_008765688.1 | 2 - 3 | 449 | 77 | |
XM_008767468.1 | XP_008765690.1 | 2 - 3 | 184 | 77 | ||
XM_008767469.1 | XP_008765691.1 | 2 - 3 | 126 | 77 | ||
XM_008767470.1 | XP_008765692.1 | 2 - 3 | 213 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
