This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01424360_g1
See other APOO GE Assays ›
Gene Symbol:
Apoo
Gene Name:
apolipoprotein O
Gene Aliases:
RGD1565289
Chromosome Location:
Chr.X: 63521033 - 63625134 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01424360_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apoo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199178.1 NP_001186107.1 3 - 4 251 84
XM_008773262.1 XP_008771484.1 3 - 4 951 84
GenBank mRNA FM122174.1 - 3 - 4 251 84
FQ210589.1 - 3 - 4 293 84
FQ218438.1 - 3 - 4 266 84
FQ221097.1 - 3 - 4 266 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00003369.rn.1 TC0X00000537.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00003368.rn.1 TC0X00000537.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0