This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01422644_m1
See other CTNNAL1 GE Assays ›
Gene Symbol:
Ctnnal1
Gene Name:
catenin (cadherin associated protein), alpha-like 1
Gene Aliases:
Catnal1
Chromosome Location:
Chr.5: 73560407 - 73621070 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01422644_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctnnal1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106649.1 NP_001100119.1 15 - 16 1988 71
XM_006238177.2 XP_006238239.1 15 - 16 3105 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002798.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
Rho protein signal transduction
cadherin binding
actin filament binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038287 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P963812 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 373927 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 194141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 581054 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 368655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 135236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278729 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 840221 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 471585 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 132965 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 967181 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 43846 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 258012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 866504 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 366705 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 891715 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 712190 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 661062 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 82685 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587353 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 387979 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 970851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 92350 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723210 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 586662 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 113598 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 144014 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 574222 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 688149 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 817417 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 178347 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 437511 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 434224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 584189 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 624330 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 945340 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 195269 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 801729 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 31662 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 88022 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 87281 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 31303 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 136745 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 17885 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P963812 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038287 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038287 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P963812 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P963812 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038287 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011664 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P963812 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80477 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 62526 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 69172 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10876910 - -
Affymetrix Rat Gene 1.0 ST Array 10876898 - -
Affymetrix Rat Gene 1.0 ST Array 10876900 - -
Affymetrix Rat Gene 1.0 ST Array 10876905 - -
Affymetrix Rat Gene 1.0 ST Array 10876909 - -
Affymetrix Rat Gene 1.0 ST Array 10876908 - -
Affymetrix Rat Gene 1.0 ST Array 10876903 - -
Affymetrix Rat Gene 1.0 ST Array 10876899 - -
Affymetrix Rat Gene 1.0 ST Array 10876912 - -
Affymetrix Rat Gene 1.0 ST Array 10876901 - -
Affymetrix Rat Gene 1.0 ST Array 10876906 - -
Affymetrix Rat Gene 1.0 ST Array 10876914 - -
Affymetrix Rat Gene 1.0 ST Array 10876911 - -
Affymetrix Rat Gene 1.0 ST Array 10876897 - -
Affymetrix Rat Gene 1.0 ST Array 10876907 - -
Affymetrix Rat Gene 1.0 ST Array 10876904 - -
Affymetrix Rat Gene 1.0 ST Array 10876902 - -
Affymetrix Rat Gene 1.0 ST Array 10876913 - -
Affymetrix Rat Gene 1.1 ST Array 10876896 - -
Rat Genome 230 2.0 Array 1381599_at - -
Rat Genome 230 2.0 Array 1398354_PM_at - -
Rat Genome 230 2.0 Array 1381599_PM_at - -
Rat Genome 230 2.0 Array 1398354_at - -
Affymetrix HT Rat Focus Array 1398354_at - -
Affymetrix HT Rat Focus Array 1398354_PM_at - -
Affymetrix Rat Clariom S Assay PSR0500020647.rn.1 TC0500002798.rn.1 RTA TC
Rat Expression Set 230 1398354_at - -
Rat Expression Set 230 1398354_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10876896 - -
Rat Genome U34 Set rc_AA946072_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21907317 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1349629 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0