This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412892_g1
See other OCM2 GE Assays ›
Gene Symbol:
Ocm2
Gene Name:
oncomodulin 2
Gene Aliases:
OM, Ocm, oncmodulin
Chromosome Location:
Chr.12: 12611105 - 12634472 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01412892_g1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ocm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012995.1 NP_037127.1 3 - 4 260 115
XM_006248852.2 XP_006248914.1 3 - 4 442 115
GenBank mRNA J02705.1 - 3 - 4 260 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000214.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ocm2

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

regulation of cytosolic calcium ion concentration
calcium ion binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158784 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15859 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 537891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 455498 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 169595 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 87473 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655905 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 462082 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224804 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 60707 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158778 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158784 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158778 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158784 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158784 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158778 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15859 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17162 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 59548 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10756669 - -
Affymetrix Rat Gene 1.0 ST Array 10756668 - -
Affymetrix Rat Gene 1.0 ST Array 10756667 - -
Affymetrix Rat Gene 1.0 ST Array 10756666 - -
Affymetrix Rat Gene 1.0 ST Array 10756670 - -
Affymetrix Rat Gene 1.1 ST Array 10756665 - -
Rat Genome 230 2.0 Array 1370477_at - -
Rat Genome 230 2.0 Array 1370477_PM_at - -
Affymetrix HT Rat Focus Array 1370477_PM_at - -
Affymetrix HT Rat Focus Array 1370477_at - -
Affymetrix Rat Clariom S Assay PSR1200001487.rn.1 TC1200000214.rn.1 RTA TC
Rat Expression Set 230 1370477_PM_at - -
Rat Expression Set 230 1370477_at - -
Affymetrix Rat Gene 1.0 ST Array 10756665 - -
Rat Genome U34 Set J02705_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22348043 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1361415 - -
RatRef-12 Expression BeadChip ILMN_61126 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0