This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412045_g1
See other NEK2L1 GE Assays ›
Gene Symbol:
Nek2l1 Nek2
Gene Name:
NIMA (never in mitosis gene a)-related kinase 2-like 1
NIMA-related kinase 2
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 15940935 - 15942589 on Build Rnor_6.0
Chromosome Location:
Chr.13: 110511546 - 110524750 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01412045_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nek2l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_345900.7 XP_345901.4 1 813 80
Gene Symbol: Nek2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053691.2 NP_446143.1 5 - 6 889 80
GenBank mRNA AF352021.1 - 5 - 6 758 80

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

mitotic sister chromatid segregation
blastocyst development
protein phosphorylation
microtubule-based process
chromosome segregation
negative regulation of DNA binding
protein autophosphorylation
centrosome separation
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
negative regulation of centriole-centriole cohesion
protein serine/threonine kinase activity
ATP binding
protein phosphatase binding
protein kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300007563.rn.1 TC1300001068.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0