This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01411620_m1
See other FLVCR1 GE Assays ›
Gene Symbol:
Flvcr1
Gene Name:
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 109578011 - 109629588 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01411620_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Flvcr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008769906.1 XP_008768128.1 4 - 5 1235 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
in utero embryonic development
mitochondrial transport
heme transport
erythrocyte differentiation
limb morphogenesis
multicellular organism growth
embryonic digit morphogenesis
regulation of organ growth
spleen development
embryonic skeletal system morphogenesis
head morphogenesis
heme export
heme transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0