This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01410126_m1
See other ATP6V0E2 GE Assays ›
Gene Symbol:
Atp6v0e2
Gene Name:
ATPase, H+ transporting V0 subunit e2
Gene Aliases:
NM9.2
Chromosome Location:
Chr.4: 78168117 - 78171265 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01410126_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp6v0e2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002253.1 NP_001002253.1 1 - 2 176 252
GenBank mRNA BC089958.1 - 128 7312897 252
CB566985.1 - 1 - 2 199 252
CB757317.1 - 1 - 2 160 252
FQ212114.1 - 1266 252

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp6v0e2

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
ATP hydrolysis coupled proton transport
proton transport
cell growth
transmembrane transport
proton-transporting ATPase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0