This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01405502_s1
See other GPR151 GE Assays ›
Gene Symbol:
Gpr151
Gene Name:
G protein-coupled receptor 151
Gene Aliases:
nGPCR-2037
Chromosome Location:
Chr.18: 36892403 - 36893797 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01405502_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr151
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181633.2 NP_853664.1 1 1277 66
GenBank mRNA AJ564168.1 - 1 1277 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr151

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P070999 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263943 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070999 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070999 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P070999 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555002 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 40738 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10804222 - -
Affymetrix Rat Gene 1.0 ST Array 10804223 - -
Affymetrix Rat Gene 1.0 ST Array 10804224 - -
Affymetrix Rat Gene 1.1 ST Array 10804221 - -
Affymetrix Rat Gene 1.0 ST Array 10804221 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22419621 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1369361 - -
RatRef-12 Expression BeadChip ILMN_49078 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0