This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01404428_m1
See other EFCAB1 GE Assays ›
Gene Symbol:
Efcab1
Gene Name:
EF hand calcium binding domain 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 90213367 - 90234286 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01404428_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Efcab1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106930.1 NP_001100400.1 2 - 3 340 130
XM_008768871.1 XP_008767093.1 2 - 3 371 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100002039.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100013628.rn.1 TC1100002039.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0