This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01403231_g1
See other PCP2 GE Assays ›
Gene Symbol:
Pcp2
Gene Name:
Purkinje cell protein 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 2172378 - 2174673 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01403231_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107116.1 NP_001100586.1 2 - 3 152 84
XM_006248762.2 XP_006248824.1 2 - 3 222 84
XM_008768953.1 XP_008767175.1 2 - 3 207 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000926.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

rhodopsin mediated signaling pathway
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200008069.rn.1 TC1200000926.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200008067.rn.1 TC1200000926.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200008068.rn.1 TC1200000926.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200008072.rn.1 TC1200000926.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0