This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01401980_m1
See other CCDC66 GE Assays ›
Gene Symbol:
Ccdc66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
RGD1560873
Chromosome Location:
Chr.16: 3106651 - 3139187 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01401980_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccdc66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008771024.1 XP_008769246.1 10 - 11 1431 73
XM_008771025.1 XP_008769247.1 10 - 11 1473 73
XM_008771026.1 XP_008769248.1 11 - 12 1557 73
XM_008771027.1 XP_008769249.1 10 - 11 1531 73
XM_008771028.1 XP_008769250.1 9 - 10 1341 73
XM_008771029.1 XP_008769251.1 10 - 11 1473 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s)

retinal rod cell development
post-embryonic retina morphogenesis in camera-type eye

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0