This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01401261_m1
See other IGFBP1 GE Assays ›
Gene Symbol:
Igfbp1
Gene Name:
insulin-like growth factor binding protein 1
Gene Aliases:
IBP1, IGF-BP25, IGFBA
Chromosome Location:
Chr.14: 87448716 - 87453783 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01401261_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igfbp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013144.1 NP_037276.1 2 - 3 717 84
GenBank mRNA BC078889.1 - 2 - 3 699 7129185 84
M58634.1 - 2 - 3 717 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400000974.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igfbp1

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
insulin receptor signaling pathway
positive regulation of cell growth
tissue regeneration
regulation of insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
insulin-like growth factor binding
insulin-like growth factor I binding
insulin-like growth factor II binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400007181.rn.1 TC1400000974.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0