This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01202378_m1
See other CABP5 GE Assays ›
Gene Symbol:
Cabp5
Gene Name:
calcium binding protein 5
Gene Aliases:
-
Chromosome Location:
Chr.1: 75693212 - 75706142 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01202378_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108907.1 NP_001102377.1 4 - 5 539 59
XM_006228328.2 XP_006228390.1 4 - 5 557 59
XM_006228329.2 XP_006228391.1 4 - 5 857 59
XM_006228330.2 XP_006228392.1 4 - 5 762 59
XM_006228331.2 XP_006228393.1 4 - 5 1629 59
XM_006228332.2 XP_006228394.1 4 - 5 1143 59
XM_006228333.2 XP_006228395.1 3 - 4 1598 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000903.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100006153.rn.1 TC0100000903.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20964652 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0