This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01176502_gH
See other SNRPA GE Assays ›
Gene Symbol:
Snrpa
Gene Name:
small nuclear ribonucleoprotein polypeptide A
Gene Aliases:
-
Chromosome Location:
Chr.1: 83999670 - 84008427 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01176502_gH
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snrpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008303.1 NP_001008304.1 6 - 7 766 67
XM_006228564.2 XP_006228626.1 6 - 7 897 67
GenBank mRNA BC086331.1 - 6 - 7 766 7300752 67
FQ227089.1 - 5 - 6 660 67
FQ231860.1 - 4 - 5 470 67
FQ233559.1 - 5 - 6 660 67

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Snrpa

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
nucleotide binding
RNA binding
snRNA binding
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P641234 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 806911 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 818179 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 563542 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 269674 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 560992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 297377 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 132509 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P641234 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P641234 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P641234 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P641234 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 72975 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10719988 - -
Affymetrix Rat Gene 1.0 ST Array 10719989 - -
Affymetrix Rat Gene 1.0 ST Array 10719984 - -
Affymetrix Rat Gene 1.0 ST Array 10719987 - -
Affymetrix Rat Gene 1.0 ST Array 10719985 - -
Affymetrix Rat Gene 1.0 ST Array 10719986 - -
Affymetrix Rat Gene 1.1 ST Array 10719983 - -
Rat Genome 230 2.0 Array 1388436_at - -
Rat Genome 230 2.0 Array 1388436_PM_at - -
Affymetrix HT Rat Focus Array 1388436_PM_at - -
Affymetrix HT Rat Focus Array 1388436_at - -
Affymetrix Rat Clariom S Assay PSR0100040035.rn.1 TC0100005033.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100040036.rn.1 TC0100005033.rn.1 RTA TC
Rat Expression Set 230 1388436_at - -
Rat Expression Set 230 1388436_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10719983 - -
Rat Genome U34 Set rc_AI230728_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21748230 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1364755 - -
RatRef-12 Expression BeadChip ILMN_60940 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0