This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01088063_g1
See other LOC501934 GE Assays ›
Gene Symbol:
LOC501934 RGD1564093
Gene Name:
uncharacterized protein C19orf43 homolog
similar to RIKEN cDNA 2310036O22
Gene Aliases:
-
-
Chromosome Location:
Chr.X: 116557457 - 116558148 on Build Rnor_6.0
Chromosome Location:
Chr.19: 26142720 - 26146136 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01088063_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC501934
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008773502.1 XP_008771724.1 2 151 64
Gene Symbol: RGD1564093
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105946.1 NP_001099416.1 2 - 3 475 64
GenBank mRNA FQ231178.1 - 2 - 3 414 64
FQ235293.1 - 2 - 3 414 64

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P106774 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044276 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 348989 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 854024 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 342232 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 176993 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044276 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106774 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044276 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106774 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044276 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P106774 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P548524 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10809996 - -
Affymetrix Rat Gene 1.0 ST Array 10809995 - -
Affymetrix Rat Gene 1.0 ST Array 10809994 - -
Affymetrix Rat Gene 1.0 ST Array 10809997 - -
Affymetrix Rat Gene 1.1 ST Array 10809993 - -
Rat Genome 230 2.0 Array 1398952_PM_at - -
Rat Genome 230 2.0 Array 1398952_at - -
Affymetrix HT Rat Focus Array 1398952_at - -
Affymetrix HT Rat Focus Array 1398952_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900002805.rn.1 TC1900000353.rn.1 RTA TC
Rat Expression Set 230 1398952_PM_at - -
Rat Expression Set 230 1398952_at - -
Affymetrix Rat Gene 1.0 ST Array 10809993 - -
Rat Genome U34 Set rc_AI233222_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21156687 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header