This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00821420_g1
See other PGAM1 GE Assays ›
Gene Symbol:
Pgam1
Gene Name:
phosphoglycerate mutase 1 (brain)
Gene Aliases:
Pgmut
Chromosome Location:
Chr.1: 261158261 - 261165808 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00821420_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053290.1 NP_445742.1 1 - 2 236 131
GenBank mRNA BC065582.1 - 1 - 2 291 5623677 131
M76591.1 - 1 - 2 236 131

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pgam1

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
regulation of glycolytic process
dephosphorylation
regulation of pentose-phosphate shunt
respiratory burst
bisphosphoglycerate mutase activity
bisphosphoglycerate 2-phosphatase activity
phosphoglycerate mutase activity
protein kinase binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12753 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12753 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12753 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12753 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12753 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P259574 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10815961 - -
Affymetrix Rat Gene 1.0 ST Array 10740239 - -
Affymetrix Rat Gene 1.0 ST Array 10940445 - -
Affymetrix Rat Gene 1.1 ST Array 10940445 - -
Affymetrix Rat Gene 1.1 ST Array 10740238 - -
Affymetrix Rat Gene 1.1 ST Array 10815960 - -
Rat Genome 230 2.0 Array 1386864_PM_at - -
Rat Genome 230 2.0 Array 1386864_at - -
Affymetrix HT Rat Focus Array 1386864_PM_at - -
Affymetrix HT Rat Focus Array 1386864_at - -
Affymetrix Rat Clariom S Assay PSR0100029480.rn.1 TC0100003622.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100029478.rn.1 TC0100003622.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100029479.rn.1 TC0100003622.rn.1 RTA TC
Rat Expression Set 230 1386864_PM_at - -
Rat Expression Set 230 1386864_at - -
Affymetrix Rat Gene 1.0 ST Array 10940445 - -
Affymetrix Rat Gene 1.0 ST Array 10815960 - -
Rat Genome U34 Set rc_AI169417_s_at - -
Rat Genome U34 Set S63233_at - -
Rat Genome U34 Set S63233_g_at - -
Rat Genome U34 Set rc_AI232001_s_at - -
Rat Genome U34 Set rc_AI228723_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21367960 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0