This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00709370_m1
See other SNAI2 GE Assays ›
Gene Symbol:
Snai2
Gene Name:
snail family zinc finger 2
Gene Aliases:
Slug
Chromosome Location:
Chr.11: 90404421 - 90406730 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00709370_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snai2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013035.1 NP_037167.1 1 - 2 83 98
GenBank mRNA AF497973.1 - 1 - 2 83 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100002040.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Snai2

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
osteoblast differentiation
epithelial to mesenchymal transition
epithelial to mesenchymal transition involved in endocardial cushion formation
cell migration involved in endocardial cushion formation
transcription from RNA polymerase II promoter
negative regulation of cell adhesion involved in substrate-bound cell migration
Notch signaling pathway
sensory perception of sound
response to radiation
negative regulation of keratinocyte proliferation
negative regulation of vitamin D biosynthetic process
neural crest cell development
cell migration
positive regulation of cell migration
negative regulation of chondrocyte differentiation
regulation of chemokine production
positive regulation of histone acetylation
negative regulation of catenin import into nucleus
desmosome disassembly
negative regulation of apoptotic process
pigmentation
negative regulation of DNA damage response, signal transduction by p53 class mediator
cellular response to fibroblast growth factor stimulus
positive regulation of fat cell differentiation
regulation of osteoblast differentiation
white fat cell differentiation
palate development
canonical Wnt signaling pathway
epithelium development
cartilage morphogenesis
regulation of branching involved in salivary gland morphogenesis
negative regulation of vitamin D receptor signaling pathway
cellular response to epidermal growth factor stimulus
cellular response to ionizing radiation
negative regulation of canonical Wnt signaling pathway
negative regulation of cell-cell adhesion by negative regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of stem cell proliferation
regulation of bicellular tight junction assembly
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100013633.rn.1 TC1100002040.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100013632.rn.1 TC1100002040.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0