This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00696783_g1
See other S100VP GE Assays ›
Gene Symbol:
S100vp
Gene Name:
S100 calcium-binding protein, ventral prostate
Gene Aliases:
S100RVP
Chromosome Location:
Chr.2: 190628525 - 190631821 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00696783_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100vp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176076.3 NP_788265.1 1 - 2 309 74
GenBank mRNA AY195741.1 - 1 - 2 313 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY195741.1, NM_176076.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100vp

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
response to testosterone
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200010917.rn.1 TC0200001625.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200010914.rn.1 TC0200001625.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200010913.rn.1 TC0200001625.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0