This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00693054_g1
See other AES GE Assays ›
Gene Symbol:
Aes
Gene Name:
amino-terminal enhancer of split
Gene Aliases:
Grg, Grg-5
Chromosome Location:
Chr.7: 11012843 - 11018156 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00693054_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aes
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019220.1 NP_062093.1 3 - 4 323 62
GenBank mRNA BC101855.1 - 288 7933368 62
L14462.1 - 3 - 4 323 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700002100.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aes

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of gene expression
Wnt signaling pathway
cellular response to extracellular stimulus
negative regulation of protein binding
regulation of growth
negative regulation of transcription, DNA-templated
negative regulation of response to cytokine stimulus
response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of anoikis
transcription corepressor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700016280.rn.1 TC0700002100.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700016281.rn.1 TC0700002100.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0