This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00691672_m1
See other PEX2 GE Assays ›
Gene Symbol:
Pex2
Gene Name:
peroxisomal biogenesis factor 2
Gene Aliases:
Pxmp3
Chromosome Location:
Chr.2: 98251725 - 98269187 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00691672_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pex2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017234.1 NP_058930.1 2 - 3 370 86
XM_006232160.2 XP_006232222.1 2 - 3 1559 86
XM_006232161.1 XP_006232223.1 2 - 3 488 86
GenBank mRNA BC063169.1 - 2 - 3 220 6886755 86
D30616.1 - 2 - 3 337 86
D30617.1 - 2 - 3 370 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC063169.1, D30616.1, D30617.1, NM_017234.1, XM_006232160.2, XM_006232161.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pex2

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
negative regulation of transcription from RNA polymerase II promoter
neuron migration
fatty acid beta-oxidation
bile acid biosynthetic process
peroxisome organization
nervous system development
biological_process
protein import into peroxisome matrix
protein destabilization
cholesterol homeostasis
regulation of cholesterol biosynthetic process
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
molecular_function
zinc ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11873 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 572234 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 515190 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 70911 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 576593 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857178 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203820 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 125116 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 87350 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 650629 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 781884 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 82361 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 369688 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391785 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 38132 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 72060 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11873 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11873 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11873 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11873 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700914427 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 37834 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 71574 - -
Agilent Rat cDNA Microarray (G4105A) 700914427 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10814382 - -
Affymetrix Rat Gene 1.0 ST Array 10814381 - -
Affymetrix Rat Gene 1.0 ST Array 10814383 - -
Affymetrix Rat Gene 1.0 ST Array 10814380 - -
Affymetrix Rat Gene 1.1 ST Array 10814379 - -
Rat Genome 230 2.0 Array 1387064_PM_at - -
Rat Genome 230 2.0 Array 1387064_at - -
Rat Genome 230 2.0 Array 1396866_PM_s_at - -
Rat Genome 230 2.0 Array 1396866_s_at - -
Affymetrix HT Rat Focus Array 1387064_at - -
Affymetrix HT Rat Focus Array 1387064_PM_at - -
Affymetrix HT Rat Focus Array 1396866_PM_s_at - -
Affymetrix HT Rat Focus Array 1396866_s_at - -
Affymetrix Rat Clariom S Assay PSR0200005001.rn.1 TC0200000814.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200005000.rn.1 TC0200000814.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200005002.rn.1 TC0200000814.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004999.rn.1 TC0200000814.rn.1 RTA TC
Rat Expression Set 230 1387064_at - -
Rat Expression Set 230 1387064_PM_at - -
Rat Expression Set 230 1396866_s_at - -
Rat Expression Set 230 1396866_PM_s_at - -
Affymetrix Rat Gene 1.0 ST Array 10814379 - -
Rat Genome U34 Set X57988mRNA_s_at - -
Rat Genome U34 Set E03344cds_s_at - -
Rat Genome U34 Set rc_AA850928_s_at - -
Rat Genome U34 Set E03344cds_s_at - -
Rat Genome U34 Set X57988mRNA_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20738260 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1366151 - -
RatRef-12 Expression BeadChip ILMN_66926 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0