This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00690481_m1
See other PLCD1 GE Assays ›
Gene Symbol:
Plcd1
Gene Name:
phospholipase C, delta 1
Gene Aliases:
Plc1
Chromosome Location:
Chr.8: 127753514 - 127782070 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00690481_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017035.1 NP_058731.1 5 - 6 1018 71
XM_006244063.2 XP_006244125.1 5 - 6 917 71
GenBank mRNA EF089258.1 - 5 - 6 796 71
M20637.1 - 5 - 6 1018 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800003537.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plcd1

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
G-protein coupled receptor signaling pathway
response to aluminum ion
response to organonitrogen compound
positive regulation of norepinephrine secretion
lipid catabolic process
positive regulation of inositol trisphosphate biosynthetic process
response to prostaglandin F
intracellular signal transduction
regulation of cell proliferation
response to peptide hormone
regulation of cytosolic calcium ion concentration
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
response to calcium ion
response to hyperoxia
labyrinthine layer blood vessel development
regulation of phospholipase C activity
phosphatidylserine binding
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
protein binding
phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
GTPase activating protein binding
phosphatidic acid binding
phosphatidylinositol phosphate binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P586141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 919651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 630616 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 859024 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 758445 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 677724 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 401923 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 858204 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 888650 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 69240 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162027 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 55399 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 93944 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P586141 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P586141 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P586141 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P586141 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 26069 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 9357 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10920818 - -
Affymetrix Rat Gene 1.0 ST Array 10920805 - -
Affymetrix Rat Gene 1.0 ST Array 10920813 - -
Affymetrix Rat Gene 1.0 ST Array 10920811 - -
Affymetrix Rat Gene 1.0 ST Array 10920814 - -
Affymetrix Rat Gene 1.0 ST Array 10920816 - -
Affymetrix Rat Gene 1.0 ST Array 10920804 - -
Affymetrix Rat Gene 1.0 ST Array 10920809 - -
Affymetrix Rat Gene 1.0 ST Array 10920806 - -
Affymetrix Rat Gene 1.0 ST Array 10920812 - -
Affymetrix Rat Gene 1.0 ST Array 10920815 - -
Affymetrix Rat Gene 1.0 ST Array 10920819 - -
Affymetrix Rat Gene 1.0 ST Array 10920807 - -
Affymetrix Rat Gene 1.0 ST Array 10920808 - -
Affymetrix Rat Gene 1.0 ST Array 10920817 - -
Affymetrix Rat Gene 1.1 ST Array 10920803 - -
Rat Genome 230 2.0 Array 1367916_PM_at - -
Rat Genome 230 2.0 Array 1367916_at - -
Affymetrix HT Rat Focus Array 1367916_PM_at - -
Affymetrix HT Rat Focus Array 1367916_at - -
Affymetrix Rat Clariom S Assay PSR0800028721.rn.1 TC0800003537.rn.1 RTA TC
Rat Expression Set 230 1367916_PM_at - -
Rat Expression Set 230 1367916_at - -
Affymetrix Rat Gene 1.0 ST Array 10920803 - -
Rat Genome U34 Set M20637_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21931641 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371243 - -
RatRef-12 Expression BeadChip ILMN_50502 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0