This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00686621_g1
See other GPR56 GE Assays ›
Gene Symbol:
Gpr56
Gene Name:
G protein-coupled receptor 56
Gene Aliases:
-
Chromosome Location:
Chr.19: 10423534 - 10460716 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00686621_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152242.2 NP_689448.2 7 - 8 1308 69
XM_006255066.2 XP_006255128.1 7 - 8 2360 69
XM_008772264.1 XP_008770486.1 7 - 8 1510 69
XM_008772265.1 XP_008770487.1 8 - 9 1774 69
XM_008772266.1 XP_008770488.1 7 - 8 3020 69
XM_008772267.1 XP_008770489.1 7 - 8 3283 69
XM_008772268.1 XP_008770490.1 8 - 9 1607 69
XM_008772269.1 XP_008770491.1 7 - 8 2389 69
XM_008772270.1 XP_008770492.1 7 - 8 2865 69
GenBank mRNA AF529886.1 - 7 - 8 1309 69
EX488169.1 - 4 - 5 419 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000985.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr56

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell adhesion
G-protein coupled receptor signaling pathway
Rho protein signal transduction
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
extracellular matrix binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1900007812.rn.1 TC1900000985.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0