This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00681074_m1
See other ADRBK2 GE Assays ›
Gene Symbol:
Adrbk2
Gene Name:
adrenergic, beta, receptor kinase 2
Gene Aliases:
Grk3
Chromosome Location:
Chr.12: 49626871 - 49750389 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00681074_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adrbk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012897.2 NP_037029.1 4 - 5 370 66
XM_008769317.1 XP_008767539.1 3 - 4 261 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000845.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adrbk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
intracellular protein transport
signal transduction
multicellular organism aging
receptor internalization
termination of G-protein coupled receptor signaling pathway
inositol phosphate metabolic process
negative regulation of G-protein coupled receptor protein signaling pathway
rhodopsin metabolic process
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
D1 dopamine receptor binding
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200007400.rn.1 TC1200000845.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0