This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00664505_g1
See other S100G GE Assays ›
Gene Symbol:
S100g
Gene Name:
S100 calcium binding protein G
Gene Aliases:
Cabp, Calb3, Cbpi, Rncalbd9
Chromosome Location:
Chr.X: 33442811 - 33445714 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00664505_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012521.2 NP_036653.1 1 - 2 52 62
XM_006256888.2 XP_006256950.1 1 - 2 427 62
GenBank mRNA BC059153.1 - 1 - 2 56 6917928 62
J02954.1 - 1 - 2 53 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC059153.1, J02954.1, NM_012521.2, XM_006256888.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100g

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
vitamin D binding
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00002387.rn.1 TC0X00000330.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00002388.rn.1 TC0X00000330.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0