This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00594854_m1
See other CRHBP GE Assays ›
Gene Symbol:
Crhbp
Gene Name:
corticotropin releasing hormone binding protein
Gene Aliases:
Crfbp
Chromosome Location:
Chr.2: 25141471 - 25153334 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00594854_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crhbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139183.2 NP_631922.1 6 - 7 930 61
GenBank mRNA X58023.1 - 6 - 7 929 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002645.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crhbp

Gene Ontology Categories:

Function(s) Process(es)

synaptic transmission, dopaminergic
maternal aggressive behavior
inflammatory response
female pregnancy
hormone-mediated signaling pathway
cellular response to drug
cellular response to potassium ion
hormone metabolic process
regulated exocytosis
behavioral response to ethanol
regulation of corticotropin secretion
negative regulation of corticotropin secretion
cellular response to calcium ion
cellular response to cocaine
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to gonadotropin-releasing hormone
negative regulation of corticotropin-releasing hormone receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
peptide binding
corticotropin-releasing hormone binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P104054 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 385863 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 231303 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 733302 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 562028 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 810487 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 387328 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291760 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 759771 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 43350 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P104054 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P104054 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P104054 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P104054 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 53844 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10820572 - -
Affymetrix Rat Gene 1.0 ST Array 10820571 - -
Affymetrix Rat Gene 1.0 ST Array 10820576 - -
Affymetrix Rat Gene 1.0 ST Array 10820570 - -
Affymetrix Rat Gene 1.0 ST Array 10820573 - -
Affymetrix Rat Gene 1.0 ST Array 10820575 - -
Affymetrix Rat Gene 1.0 ST Array 10820574 - -
Affymetrix Rat Gene 1.1 ST Array 10820569 - -
Rat Genome 230 2.0 Array 1369908_at - -
Rat Genome 230 2.0 Array 1369908_PM_at - -
Affymetrix HT Rat Focus Array 1369908_PM_at - -
Affymetrix HT Rat Focus Array 1369908_at - -
Affymetrix Rat Clariom S Assay PSR0200018498.rn.1 TC0200002645.rn.1 RTA TC
Rat Expression Set 230 1369908_PM_at - -
Rat Expression Set 230 1369908_at - -
Affymetrix Rat Gene 1.0 ST Array 10820569 - -
Rat Genome U34 Set AF003904mRNA_g_at - -
Rat Genome U34 Set AF003904mRNA_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21537974 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1355385 - -
RatRef-12 Expression BeadChip ILMN_59075 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0