This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00592428_m1
See other SERPINA10 GE Assays ›
Gene Symbol:
Serpina10 LOC100909524
Gene Name:
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
protein Z-dependent protease inhibitor-like
Gene Aliases:
Rasp1
-
Chromosome Location:
Chr.6: 127500014 - 127508470 on Build Rnor_6.0
Chromosome Location:
Chr.6: 129004249 - 129012705 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn00592428_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serpina10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133617.2 NP_598301.2 3 - 4 1096 76
GenBank mRNA BC088113.1 - 3 - 4 1096 7326099 76
U55765.1 - 2 - 3 1008 76
Gene Symbol: LOC100909524
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003750227.3 XP_003750275.1 3 - 4 1114 76

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serpina10
Click here to find siRNAs for Gene LOC100909524

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
negative regulation of endopeptidase activity
tissue regeneration
regulation of acrosome reaction
serine-type endopeptidase inhibitor activity
heparin binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P083369 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171891 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 64520 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 417716 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 553303 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 182879 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 461828 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 12512 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 71047 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083369 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083369 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P083369 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543096 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 35966 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 51191 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10891919 - -
Affymetrix Rat Gene 1.0 ST Array 10891918 - -
Affymetrix Rat Gene 1.0 ST Array 10891917 - -
Affymetrix Rat Gene 1.0 ST Array 10891921 - -
Affymetrix Rat Gene 1.0 ST Array 10891920 - -
Affymetrix Rat Gene 1.1 ST Array 10891916 - -
Rat Genome 230 2.0 Array 1368790_PM_at - -
Rat Genome 230 2.0 Array 1368790_at - -
Affymetrix HT Rat Focus Array 1368790_at - -
Affymetrix HT Rat Focus Array 1368790_PM_at - -
Affymetrix Rat Clariom S Assay PSR0600021852.rn.1 TC0600003070.rn.1 RTA TC
Rat Expression Set 230 1368790_PM_at - -
Rat Expression Set 230 1368790_at - -
Affymetrix Rat Gene 1.0 ST Array 10891916 - -
Rat Genome U34 Set U55765_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21846151 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1354651 - -
RatRef-12 Expression BeadChip ILMN_53532 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0