This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00582568_m1
See other GPR6 GE Assays ›
Gene Symbol:
Gpr6
Gene Name:
G protein-coupled receptor 6
Gene Aliases:
-
Chromosome Location:
Chr.20: 45813169 - 45815940 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00582568_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031806.1 NP_113994.1 1 - 2 132 76
GenBank mRNA AF064706.1 - 1 - 2 132 76
U12006.1 - 1 - 2 132 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF064706.1, NM_031806.1, U12006.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr6

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

sphingosine-1-phosphate signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
sphingosine-1-phosphate receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000013758.rn.1 TC2000001697.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000013757.rn.1 TC2000001697.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0