This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00575407_s1
See other ADNP GE Assays ›
Gene Symbol:
Adnp
Gene Name:
activity-dependent neuroprotector homeobox
Gene Aliases:
-
Chromosome Location:
Chr.3: 164937188 - 164964819 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00575407_s1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adnp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022681.2 NP_073172.2 4 2335 96
GenBank mRNA AF234680.1 - 1 1355 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adnp

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
short-term memory
response to carbohydrate
response to inorganic substance
negative regulation of gene expression
regulation of protein ADP-ribosylation
positive regulation of neuron projection development
positive regulation of cGMP biosynthetic process
cellular response to extracellular stimulus
negative regulation of protein binding
activation of protein kinase activity
nitric oxide homeostasis
negative regulation of neuron apoptotic process
estrous cycle
positive regulation of axon extension
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of synaptic transmission
positive regulation of synapse assembly
DNA binding
chromatin binding
copper ion binding
peptide binding
metal ion binding
beta-tubulin binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P634358 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 102463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716712 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 261347 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 801643 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 217373 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 935368 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325243 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 42768 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 132975 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634358 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634358 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P634358 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P634358 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P843366 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 95905 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 34928 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10851993 - -
Affymetrix Rat Gene 1.0 ST Array 10851994 - -
Affymetrix Rat Gene 1.0 ST Array 10851996 - -
Affymetrix Rat Gene 1.0 ST Array 10851995 - -
Affymetrix Rat Gene 1.0 ST Array 10851992 - -
Affymetrix Rat Gene 1.1 ST Array 10851991 - -
Rat Genome 230 2.0 Array 1389088_at - -
Rat Genome 230 2.0 Array 1389346_PM_at - -
Rat Genome 230 2.0 Array 1389346_at - -
Rat Genome 230 2.0 Array 1392456_PM_at - -
Rat Genome 230 2.0 Array 1389088_PM_at - -
Rat Genome 230 2.0 Array 1369402_at - -
Rat Genome 230 2.0 Array 1369402_PM_at - -
Rat Genome 230 2.0 Array 1392456_at - -
Affymetrix HT Rat Focus Array 1389346_at - -
Affymetrix HT Rat Focus Array 1369402_PM_at - -
Affymetrix HT Rat Focus Array 1389088_at - -
Affymetrix HT Rat Focus Array 1392456_at - -
Affymetrix HT Rat Focus Array 1369402_at - -
Affymetrix HT Rat Focus Array 1389346_PM_at - -
Affymetrix HT Rat Focus Array 1389088_PM_at - -
Affymetrix HT Rat Focus Array 1392456_PM_at - -
Rat Expression Set 230 1389088_at - -
Rat Expression Set 230 1392456_at - -
Rat Expression Set 230 1369402_at - -
Rat Expression Set 230 1389088_PM_at - -
Rat Expression Set 230 1389346_at - -
Rat Expression Set 230 1392456_PM_at - -
Rat Expression Set 230 1369402_PM_at - -
Rat Expression Set 230 1389346_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10851991 - -
Rat Genome U34 Set rc_AA874881_at - -
Rat Genome U34 Set rc_AA900236_at - -
Rat Genome U34 Set rc_AI232565_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21503181 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360977 - -
RatRef-12 Expression BeadChip ILMN_65360 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0