This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00573666_s1
See other HTR1B GE Assays ›
Gene Symbol:
Htr1b
Gene Name:
5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled
Gene Aliases:
-
Chromosome Location:
Chr.8: 89113997 - 89130829 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00573666_s1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Htr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022225.1 NP_071561.1 1 656 91
GenBank mRNA X62944.1 - 1 692 91

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Htr1b

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor internalization
adenylate cyclase-inhibiting serotonin receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
serotonin receptor signaling pathway
chemical synaptic transmission
feeding behavior
response to toxic substance
regulation of dopamine secretion
negative regulation of serotonin secretion
negative regulation of cAMP biosynthetic process
negative regulation of synaptic transmission, GABAergic
cellular response to drug
response to cocaine
vasoconstriction
response to drug
drinking behavior
response to ethanol
bone remodeling
regulation of behavior
response to mineralocorticoid
negative regulation of synaptic transmission, glutamatergic
cellular response to alkaloid
cellular response to temperature stimulus
G-protein coupled serotonin receptor activity
drug binding
serotonin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800024155.rn.1 TC0800003019.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0