This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00568246_m1
See other HSD17B7 GE Assays ›
Gene Symbol:
Hsd17b7
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 7
Gene Aliases:
-
Chromosome Location:
Chr.13: 88288116 - 88308011 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00568246_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd17b7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017235.3 NP_058931.1 1 - 2 105 64
XM_006250215.2 XP_006250277.1 1 - 2 128 64
XM_006250216.2 XP_006250278.1 1 - 2 128 64
XM_006250217.2 XP_006250279.1 1 - 2 128 64
GenBank mRNA FM073997.1 - 1 - 2 98 64
FQ113345.1 - 1 - 2 105 64
U44803.1 - 1 - 2 50 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001874.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd17b7

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
estrogen biosynthetic process
oxidation-reduction process
3-keto sterol reductase activity
estradiol 17-beta-dehydrogenase activity
prolactin receptor binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093269 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 888637 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 647279 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 957611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 477920 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 940398 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 242224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 640287 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 825983 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 625803 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 662823 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325883 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 665216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 869755 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 73807 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 95919 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093269 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093269 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093269 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P796502 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 702025708 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700935795 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700935203 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 22720 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 42822 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 34461 - -
Agilent Rat cDNA Microarray (G4105A) 700935795 - -
Agilent Rat cDNA Microarray (G4105A) 700935203 - -
Agilent Rat cDNA Microarray (G4105A) 702025708 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10769690 - -
Affymetrix Rat Gene 1.0 ST Array 10769686 - -
Affymetrix Rat Gene 1.0 ST Array 10769684 - -
Affymetrix Rat Gene 1.0 ST Array 10769687 - -
Affymetrix Rat Gene 1.0 ST Array 10769683 - -
Affymetrix Rat Gene 1.0 ST Array 10769685 - -
Affymetrix Rat Gene 1.0 ST Array 10769682 - -
Affymetrix Rat Gene 1.0 ST Array 10765456 - -
Affymetrix Rat Gene 1.0 ST Array 10769688 - -
Affymetrix Rat Gene 1.0 ST Array 10769691 - -
Affymetrix Rat Gene 1.0 ST Array 10765455 - -
Affymetrix Rat Gene 1.0 ST Array 10769681 - -
Affymetrix Rat Gene 1.0 ST Array 10769689 - -
Affymetrix Rat Gene 1.1 ST Array 10765452 - -
Affymetrix Rat Gene 1.1 ST Array 10769680 - -
Rat Genome 230 2.0 Array 1387233_at - -
Rat Genome 230 2.0 Array 1387233_PM_at - -
Affymetrix HT Rat Focus Array 1387233_PM_at - -
Affymetrix HT Rat Focus Array 1387233_at - -
Affymetrix Rat Clariom S Assay PSR1300013246.rn.1 TC1300001874.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300013247.rn.1 TC1300001874.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300013248.rn.1 TC1300001874.rn.1 RTA TC
Rat Expression Set 230 1387233_at - -
Rat Expression Set 230 1387233_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10769680 - -
Rat Genome U34 Set rc_AI236145_at - -
Rat Genome U34 Set rc_AA850737_g_at - -
Rat Genome U34 Set rc_AA850737_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21290371 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1361546 - -
RatRef-12 Expression BeadChip ILMN_67645 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0