This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00561700_m1
See other S100A4 GE Assays ›
Gene Symbol:
S100a4
Gene Name:
S100 calcium-binding protein A4
Gene Aliases:
18A2, 42A, CAPL, MTS1, P9ka, PEL98, RNP9KA
Chromosome Location:
Chr.2: 189997278 - 189999587 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00561700_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012618.2 NP_036750.1 1 - 2 37 62
GenBank mRNA FQ215053.1 - 1 - 2 37 62
FQ220722.1 - 1 - 2 37 62
FQ221062.1 - 1 - 2 37 62
FQ221355.1 - 1 - 2 37 62
FQ221406.1 - 1 - 2 38 62
FQ221428.1 - 1 - 2 37 62
FQ221699.1 - 1 - 2 40 62
FQ221706.1 - 1 - 2 38 62
FQ221757.1 - 1 - 2 38 62
FQ221860.1 - 1 - 2 38 62
FQ222027.1 - 1 - 2 38 62
FQ222069.1 - 1 - 2 37 62
FQ222219.1 - 1 - 2 38 62
FQ222585.1 - 1 - 2 37 62
FQ222875.1 - 1 - 2 37 62
FQ222962.1 - 1 - 2 38 62
FQ223047.1 - 1 - 2 41 62
FQ223128.1 - 1 - 2 38 62
FQ223234.1 - 1 - 2 37 62
FQ228364.1 - 1 - 2 39 62
FQ228506.1 - 1 - 2 37 62
FQ228607.1 - 1 - 2 39 62
FQ229874.1 - 1 - 2 37 62
J03628.1 - 1 - 2 38 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the J03628.1, NM_012618.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Rat Tumor Metastasis Panel Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a4

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
calcium ion binding
protein binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding
protein heterodimerization activity
calcium-dependent protein binding
RAGE receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200010862.rn.1 TC0200001618.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200010864.rn.1 TC0200001618.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0